Description
Many biopharmaceutical formulations use carbohydrates to enhance the stability of proteins. This method is an example of a "heart-cut" 2D HPLC method, separating the carbohydrate analyte from the protein. The method uses two charge aerosol detectors, as an example, but one can be substituted by UV. Calibration for the Trehalose is from 15.6 - 2000 ng o.c. If a second Corona Veo SD is used for the Protein, it uses the parameters described as "Veo SD".| Market: | Other |
| Keywords: | carbohydrate, Protein, BSA, Bovine serum albumin, trehalose |
| Matrix: | Water |
| Author: | Marc Plante |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.