Find methods for your needs
Consolidated GC-MS/MS Analysis of OCPs, PAHs, and PCBs in Environmental Samples
Description
This application describes the analysis of semivolatile compounds like Organochlorine pesticides (OCP), polyaromatic hydrocarbons (PAH), and polychlorinated biphenyls (PCB) in various environmental matrices, showing the productivity and high quality results of a consolidated GC-MS/MS method using the Thermo Scientific™ TRACE™ 1310 GC and the TSQ™ 8000 triple quadrupole GC-MS/MS. It also describes the use of smart software tools that are integrated into the method development and analysis workflow to minimize the time needed to implement and maintain the methodology in routine.| Market: | Environmental |
| Keywords: | Organochlorine pesticides, Polyaromatic hydrocarbons, Polychlorinated biphenyls, building material, soil samples, water samples |
| Matrix: | Soil, Water |
| Author: | Inge de Dobbeleer, Joachim Gummersbach, Hans-Joachim Huebschmann, Anton Mayer, Paul Silcock |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.