Find methods for your needs
CAN101: Detection of Ethyl Glucuronide Using Pulsed Electrochemical Detection following Reversed Phase High Performance Liquid Chromatography
Description
This application note discusses improvements made to the previous method. One change involves mobile phase optimization, in which alternative organic modifiers are studied. Another change is the addition of a wash step, which prevents carryover of the sample matrix between runs. Adjustments made to the SPE provide a much higher sample recovery. The internal standard is also changed to ProG in order to prevent elution with matrix interferents.| Market: | Clinical Research; Pharma |
| Keywords: | Acclaim PolarAdvantage, CAN101, EtG, Ethanol Metabolism, PED |
| Matrix: | Water |
| Author: | Ivy Grimm, Romina Shah, William R. LaCourse |
| Affiliation: | Department of Chemistry and Biochemistry, University of Maryland, Baltimore County |
For Research Use Only. Not for use in diagnostic procedures.