Find methods for your needs
TN50: Determination of the Amino Acid Content of Peptides by AAA-Direct
Description
This Technical Note describes the peptide hydrolysis procedures that lead to successful determinations of amino acid composition using AAA-Direct. Results for the amino analysis of two peptides, Melanocyte Stimulating Hormone (MSH) and Luteinizing Hormone-Releasing Hormone (LH-RH), using both hydrochloric acid and methanesulfonic acid (MSA) as hydrolyzing reagents are presented to demonstrate the performance of this method.Market: | BioPharma; Food and Beverage |
Keywords: | AminoPac PA10, AAA-Direct gold electrode, LH-RH, MSH, Ninhydrin, TN50, IonPac ATC-3 |
Matrix: | DI water |
Author: | Valoran Hanko and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.