Find methods for your needs
Separation and determination of acesulfame K and sucralose in beer using a polar embedded HPLC column with RI detection
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation and determination of acesulfame K and sucralose in beer. The separation was performed on a Thermo Scientific Acclaim Polar Advantage II (PA2) HPLC column. The results obtained for resolution between acesulfame K and sucralose was greater than 5 and tailing factor were less than 2.0 for both compounds.| Market: | Food and Beverage |
| Keywords: | HPLC, Sucralose, RI detection, Beer, Acclaim Polar Advantage II (PA2), Acesulfame K |
| Matrix: | Beer |
| Author: | Krishna Rao Dara |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.