Find methods for your needs
AB140: Fast Determination of Inorganic Counterions in a Pharmaceutical Drug Using High-Pressure Capillary Ion Chromatography
Description
This application brief describes the determination of counter anion chloride in a drug for type 2 diabetes using the Thermo Scientific Dionex ICS-5000 capillary IC system and Dionex IonSwift MAX-100 capillary column. The Eluent Generation Cartridge lasts 18 months under this condition in continuous operation mode and each analysis is less than 8 min.| Market: | Pharma |
| Keywords: | counter ion, anions, RFIC, ion chromatography, Chloride, Anion-exchange, Suppressed Conductivity, capillary IC, Drug, Dionex IonSwift MAX-100 column |
| Matrix: | Drug |
For Research Use Only. Not for use in diagnostic procedures.