Description
Bromate is separated using a latexbased anion exchanger and a methanesulfonic acid eluent, followed by a simplified postcolumn reaction to form triiodide. This is subsequently detected by its UV-absorption. Because the reaction takes place at lower pH than other methods, no heating of the reaction coil is needed. These conditions prevent interference from chlorite, which is known to interfere in other bromate determination methods.| Market: | Environmental |
| Keywords: | CarboPac PA1 column, Acidic Eluent, Ion Chromatography, Ozonation, Triiodide, UV-absorption Detection |
| Matrix: | Water |
| Author: | Jules Djoukeng and Detlef Jensen |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.