Find methods for your needs
AN1145: Determination of Halogens in Coal Using Combustion Ion Chromatography
Description
When coal is burned, the exhaust contains halogens that can be released into the atmosphere as acid gases such as hydrogen chloride (HCl) and hydrogen fluoride (HF), which can harm vegetation, animals, and infrastructure directly or cause acid rain. In this application note, coal samples were combusted in the Mitsubishi AQF-2100H system and then fluorine and chlorine were accurately and precisely determined using the Dionex ICS-2100 Ion Chromatography System with a Dionex IonPac AS15 anion-exchange column set.| Market: | Environmental |
| Keywords: | IonPac AS15, Pollution, power plant, RFIC, Acid rain, CIC, Combustion, AQF-2100H, halogens, coal, TN000767 |
| Matrix: | Coal |
| Author: | Adelon Agustin1, Carl Fisher2, and Mark Manahan1 |
| Affiliation: | 1-COSA Xentaur, Yaphank, NY, USA 2-Thermo Fisher Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.