Find methods for your needs
Determination of Chloride in Algal Oil Using Combustion Ion Chromatography
Description
Oil extracted from fish and algae is a renewable alternative to petroleum-based sources. If this oil is used as a lubricant, chloride needs to be monitored and its level minimized to reduce the likelihood that corrosive HCl will form. By using a combustion ion chromatography (CIC) system consisting of the automatic quick furnace AQF-2100H and an ICS-2100 ion chromatograph, the concentration of chloride can be determined accurately and quickly.| Market: | Petrochem, Oil and Gas |
| Keywords: | IonPac AS15, Renewable Energy, RFIC, Combustion, Corrosive, AQF-2100H, Fish oil, Algae, Lubricant, TN000767 |
| Matrix: | Algal oil |
For Research Use Only. Not for use in diagnostic procedures.