Find methods for your needs
Determination of Halogens and Sulfur in a Polyethylene Standard Reference Material by Combustion Ion Chromatography
Description
Polyethylene (PE) is widely used for containers, packaging material, etc. Because halogen compounds are often used as additives, it is important to know the halogen content when disposing or recycling PE products. By using a combustion ion chromatography (CIC) system consisting of the automatic quick furnace AQF-2100H and an ICS-1600 ion chromatograph, the concentrations of halogens and sulfur can be determined accurately and quickly.| Market: | Environmental |
| Keywords: | Additives, Packaging, IonPac AS12A, Sulfur, Chlorine, AQF-2100H, halogens, plastic, Polyethylene, Bromine, Reference material, recycling, TN000767 |
| Matrix: | Polyethylene |
For Research Use Only. Not for use in diagnostic procedures.