Find methods for your needs
Determination of Chlorophenols in Water according to US EPA Method 528
Description
The application provides a complete analytical workflow for the GC-MS analysis of phenolic compounds in water according to the EPA 528 guideline. The method includes all the steps required to collect, prepare and analyze samples. The extraction and purification of the analytes have been achieved with the Thermo Scientific Dionex AutoTrace 280 SPE for complete unattended operations.| Market: | Environmental |
| Keywords: | AutoTrace 280 SPE, Chlorophenols, Chromeleon CDS, EPA 528, Solid phase extraction, Water Pollutants, Water Quality |
| Matrix: | Water |
| Author: | Andrea Caruso;Tommaso Albertini;Fabrizio Galbiati |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.