Find methods for your needs
Analysis of BTEX and Chlorinated Compounds in Water via a Dual Detector Configuration Gas Chromatograph
Description
The application describes the determination of water pollutants as volatile aromatics and chlorinated hydrocarbons through a dual detector configuration of the Trace1310 GC, using the FID for the quantitation of the aromatic hydrocarbons and a more selective ECD for the quantitation of the halogenated compounds. A double detector configuration allows the simultaneous analysis of both classes of compounds, thanks to a post-column splitting of the effluents to the two detectors. A dedicated Dual Detector Kit has been used to guarantee inertness of the sample path and no dead volumes.| Market: | Environmental |
| Keywords: | BTEX, Chlorinated Hydrocarbons, Chromeleon CDS, Dual-Detector, Static Headspace, Trace1310 GC, Triplus300 HS, Water Pollutants |
| Matrix: | Water |
| Author: | Andrea Caruso |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.