Find methods for your needs
AU148: Determination of Perchlorate in Drinking Water Using a Reagent-Free Ion Chromatography System (Part B)
Description
This Application Update updates the EPA Method 314.0 for perchlorate determination in drinking water with an improved suppressor, the Dionex™ AERS 500 suppressor. The Dionex AERS 500 suppressor is a superior suppressor as it provides high capacity, low noise, high backpressure resilience, and fast startup. Moreover, the AERS 500 suppressor minimizes peak dispersion and improves peak efficiency. This document evaluates and describes the linear range and initial demonstration of capability (EPA Method 314.0, Section 9.2). (This part has a one-click workflow using ICS5000 and AS-DV autosampler).| Market: | Water Analysis |
| Keywords: | Drinking Water, RFIC, IonPac AS16, CCL, AU 148, EPA Method 314.0, MCT Verification |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.