Find methods for your needs
AU148: Determination of Perchlorate in Drinking Water Using Reagent-Free™ Ion Chromatography
Description
This application update describes the determination of perchlorate using the procedure outlined in EPA Method 314.0. This application used an integrated ion chromatography system with a 4 mm Dionex IonPac AS16 column, a Thermo Scientific™ Dionex™ EluGen™ EGC II KOH cartridge, a 1000 µL injection, and suppressed conductivity detection with a Dionex ASRS ULTRA II operated in external water mode.| Market: | Water Analysis |
| Keywords: | Drinking Water, CCL, AU 148, EPA Method 314.0, IonPac AS16, RFIC |
For Research Use Only. Not for use in diagnostic procedures.