Find methods for your needs
AN1007: Determination of Mono-, Di-, and Triphosphates and Citrate in Shrimp by Ion Chromatography
Description
This work shows an accurate IC method for determination of mono-, di-, and triphosphate species and citrate in shrimp. The sample is treated on-line using the Dionex InGuard HRP cartridge to reduce sample preparation time, error, and contamination. The eluent used in this application is produced by an eluent generator to preclude labor or error associated with eluent preparation. Overall, the method is labor efficient and accurate.| Market: | Food and Beverage |
| Keywords: | AN 1007, Dionex InGuard HRP cartridge, IonPac AS11, RFIC, Seafood |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.