Find methods for your needs
Analysis of perfluoro-octanoic acid (PFOA) and perfluorooctanesulfonic acid (PFOS) in tap water using reversed-phase HPLC with suppressed conductivity detection
Description
The separation of perfluoro-octanoic acid (PFOA) and perfluorooctanesulfonic acid (PFOS) in tap water was performed on a Thermo Scientific Acclaim PolarAdvantage II (PA2) HPLC column, with on-line sample concentration and suppressed conductivity detection. Chromatogram A is Sunnyvale tap water spiked with PFOA and PFOS and chromatogram B is a control.| Market: | Environmental |
| Keywords: | Acclaim PA2, Acclaim PolarAdvantage II, HPLC, Perfluorooctanesulfonic acid, Perfluoro-octanoic acid, PFOA, PFOS, Suppressed conductivity detection, Tap water |
| Matrix: | Tap water |
| Author: | Mark Tracy, Xiaodong Liu, Christopher Pohl |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.