Description
Tween 80, or polysorbate 80, is a polymeric, non-ionic surfactant. Due to the variety of different structures contained within this complex product, a characterization method is described to determine the relative amounts of the different components, which may vary from lot-to-lot.| Market: | BioPharma |
| Keywords: | Tween 80, Polysorbate 80 |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.