Find methods for your needs
Fast analysis of triclosan in toothpaste using a Thermo Scientific Acclaim RSLC C8 column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of triclosan in toothpaste with minimal sample preparation. The separation is performed on a Thermo Scientific Acclaim RSLC C8 column, with UV detection at 281 nm. Sample preparation: 1.0 g Toothpaste + 1.0 mL of 7.5 mg/mL magnesium sulfate + methanol to make 25 mL. Sonicate and filter.| Market: | Other |
| Keywords: | Acclaim C8, HPLC, Saccharin, Toothpaste, Triclosan, UV detection |
| Matrix: | Toothpaste |
For Research Use Only. Not for use in diagnostic procedures.