Find methods for your needs
USP 467 analysis of residual solvents using GC-FID with headspace
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of residual solvents by USP method 467. The separation was performed on a Thermo Scientific TRACE TR-V1 GC column, with flame ionization detection.| Market: | Chemical |
| Keywords: | Acetonitrile, Benzene, Carbon tetrachloride, Chlorobenzene, Cyclohexane, Dichloroethane, Dichloromethane, Dioxane, Ethyl benzene, FID, GC, Headspace, Methanol, Methylcyclohexane, Residual solvents, Tetrahydrofuran, Toluene, TRACE TR-V1, Trichloroethane, USP 467, Xylene |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.