Find methods for your needs
Fast and Easy HPLC Method Development: Application of an Automated Method Scouting System for the Separation of Hypertension Drug Formulation
Description
The Automated Method Scouting Solution with Chromeleon CDS 7.2 allows an efficient screening of methods. The user-friendly solution requires no manual interaction and runs unattended. By applying the Custom Injection Variables the needed instrument methods for altering chromatographic conditions is decreased to a minimum. Easy generation of a method scouting sequence is provided by a dedicated eWorkflow which already contains Custom Injection Variables and related instrument methods. Predefined test case criteria and queries help to extract the most promising methods.| Market: | Pharma |
| Keywords: | Column Switching, Custom Injection Variables, Intelligent Run Control, Queries, Spectral Library, Solvent Selection Valve |
| Matrix: | N/A |
| Author: | Susanne Fabel |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.