Find methods for your needs
Simple gradient method for the analysis of protein standards by micro LC
Description
The Thermo Scientific HPLC system is applied for the analysis of protein standards. The separation is performed on a Thermo Scientific Acclaim 300 C18 micro LC column, with UV detection at 214 nm.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, Cytochrome C, HPLC, Insulin, Micro LC, Myoglobin, Protein, Ribonuclease, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.