Find methods for your needs
Comparison of two reversed-phase columns for the analysis of tryptic digests of proteins
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of tryptic digests of proteins. The separation is performed on Thermo Scientific Acclaim RSLC C18 and Acclaim RSLC PolarAdvantage II 2.2µm columns, with UV detection at 210 nm.| Market: | BioPharma |
| Keywords: | Acclaim RSLC C18, Acclaim RSLC PolarAdvantage II, Bovine serum albumin, HPLC, Protein, Tryptic digest, UV detection |
| Matrix: | Bovine serum albumin |
For Research Use Only. Not for use in diagnostic procedures.