Find methods for your needs
Fast and simple isocratic method for the analysis of vitamin K1 and K2
Description
The Thermo Scientific HPLC system is applied for the analysis of vitamin K1 and K2. The separation is performed on a Thermo Scientific Acclaim HILIC-10 column, with UV detection at 265 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim HILIC-10, Fat-soluble vitamin, HILIC, HPLC, UV detection, Vitamin K1, Vitamin K2, Vitamins |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.