Find methods for your needs
Fast isocratic separation of six cardiac antiarrhythmic drugs (beta-blockers)
Description
The Thermo Scientific HPLC system is applied for the analysis of beta-blockers. The separation is performed using a Thermo Scientific Acclaim 120 C8 column, with UV detection at 214 nm.Market: | Pharma |
Keywords: | Acclaim 120 C8, Acebutolol, Antiarrhythmic, Beta-blockes, Cardiac, HPLC, Labetalol, Maleic acid, Metoprolol, Oxprenolol, Propranolol, Timolol, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.