Description
The Thermo Scientific HPLC system is applied for the analysis of sulfa drugs. The separation is performed using a Thermo Scientific Acclaim PolarAdvantage column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage, HPLC, Sulfa drugs, Sulfadiazine, Sulfadimethoxine, Sulfamerazine, Sulfamethazine, Sulfamethoxazole, Sulfanilamide, Sulfanilic acid, Sulfathiazole, Sulfisoxazole, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.