Find methods for your needs
AN106: Ion Chromatography in the Pharmaceutical Industry (Anions)
Description
Ion Chromatography is a simple and accurate technique for the analysis and quantification of charged species. This application note describes the use of ion exchange chromatography and ion exclusion chromatography in the pharmaceutical industry for the analysis of ions in ingredients and final products. The topic of cleaning validation is also discussed as it applies to charged species. Here shows the separation of 8 anions that might be found in pharmaceutical formulations by anion exchange chromatography using the IonPac AS11 column.| Market: | Pharma |
| Keywords: | Bromide, Chloride, Citrate, counterions, Dionex IonPac AS11 column, Fluoride, IC, ion chromatography, Maleate, Nitrite, Phosphate, Sulfate |
| Matrix: | Drug |
For Research Use Only. Not for use in diagnostic procedures.