Find methods for your needs
Comparison of five reversed-phase columns for the separation of 4-chloro, 7-nitro, 1,2,3-benzoxadiazole (NBD-chloride) derivatives of volatile amines
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of 4-chloro, 7-nitro, 1,2,3-benzoxadiazole (NBD-chloride) derivatives of volatile amines. The separation is performed on five reversed-phase columns; Thermo Scientific Acclaim C30, Acclaim 120 C8, Acclaim 120 C18, Acclaim PolarAdvanage and Acclaim PolarAdvantage II. The results were compared.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, Acclaim 120 C8, Acclaim C30, Acclaim PolarAdvanage, Acclaim PolarAdvantage II, HPLC, NBD derivatives, NBD-diethylamine, NBD-dimethylamine, NBD-ethylamine, NBD-methylamine, UV detection, Volatile amines |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.