Find methods for your needs
AN134: Determination of Low Concentrations of Perchlorate in Drinking and Ground Waters Using Ion Chromatography
Description
Perchlorate, a human health concern, has been found in drinking water wells in regions of the U.S. where aerospace material, munitions, or fireworks were developed, tested, or manufactured. This application note shows that perchlorate can be quantified at the 2 μg/L level using a Dionex IonPac AS16 column, hydroxide eluent generation, a large-loop injection, and suppressed conductivity detection. Refer to AU148 for a description of how detection levels of 1 μg/L or less can be obtained using an improved suppressor.| Market: | Environmental |
| Keywords: | Drinking Water, IonPac AS16, RFIC, Brine, Perchlorate, Ammonium Perchlorate, OnGuard Ag, OnGuard H, Ground water |
| Matrix: | Water |
| Author: | Swati Gokhale, Peter Jackson, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.