Find methods for your needs
Enhanced resolution of polyunsaturated lipids in a nutritional supplement by HPLC-CAD
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of polyunsaturated lipids in a nutritional supplement. The separation is performed on a Thermo Scientific Accucore C30 HPLC column, with charged aerosol detection (CAD).| Market: | Food and Beverage |
| Keywords: | Lipid, Accucore C30, ALA, CAD, Charged aerosol detection, DHA, EPA, GLA, HPLC, Nutritional supplement, Oil, Polyunsaturated lipids, Fatty acid |
| Matrix: | Oil |
For Research Use Only. Not for use in diagnostic procedures.