Find methods for your needs
Simple assay analysis of norfloxacin and metronidazole in formulation using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the assay analysis of norfloxacin and metronidazole in formulation using a simple reversed-phase isocratic method. The two compounds were separated using a Thermo Scientific Hypersil GOLD 5 µm HPLC column in less than 10 minutes with UV detection at 290 nm.| Market: | Pharma |
| Keywords: | Antibiotic, HPLC, Norfloxacin, UV detection, Metronidazole |
| Matrix: | Formulation |
For Research Use Only. Not for use in diagnostic procedures.