![](/Content/Images/icons/rate.png?v=2511129)
![](/Content/Images/icons/ask_expert.png?v=2511129)
![](/Content/Images/icons/request.png?v=2511129)
![](/Content/Images/icons/favorites.png?v=2511129)
![](/Content/Images/icons/create_parts_list.png?v=2511129)
Parts List
![](/Content/Images/icons/send_email.png)
Friend
![](/Content/Images/icons/print.png?v=2511129)
Find methods for your needs
![](/Content/Images/icons/search_help.png?v=2511129)
AN131: Determination of Transition Metals at PPT Levels in High-Purity Water and SC2 (D-clean) Baths
Description
Semiconductor cleaning solutions used to remove potentially damaging transition metal contamination are monitored to assess cleaning efficiency. Here trace concentrations (ng/L) of transition metals in semiconductor cleaning solutions and deionized water are separated using the strong complexing agent, pyridine-2,6-dicarboxylic acid (PDCA), on an IonPac CS5A column optimized with anion- and cation-exchange chemistry. Then a post-column derivitization agent, 4-(2- pyridilazo)resorcinol (PAR), is added to displace PDCA and to facilitate detection at 520–530 nm.Market: | Water Analysis |
Keywords: | copper, Iron, PAR, PDCA, Zinc, absorbance, IonPac CS5A, post column, semiconductor cleaning bath, t-metals, Transition Metals, Visible Absorbance, semiconductor, Nickel, TN 73892, TN73892, AN 131 |
Matrix: | Semiconductor cleaning bath |
Author: | Mark Laikthman and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.