Find methods for your needs
AN131: Determination of Transition Metals at PPT Levels in High-Purity Water and SC2 (D-clean) Baths
Description
Semiconductor cleaning solutions used to remove potentially damaging transition metal contamination are monitored to assess cleaning efficiency. Here trace concentrations (ng/L) of transition metals in semiconductor cleaning solutions and deionized water are separated using the strong complexing agent, pyridine-2,6-dicarboxylic acid (PDCA), on an IonPac CS5A column optimized with anion- and cation-exchange chemistry. Then a post-column derivitization agent, 4-(2- pyridilazo)resorcinol (PAR), is added to displace PDCA and to facilitate detection at 520–530 nm.| Market: | Water Analysis |
| Keywords: | copper, Iron, PAR, PDCA, Zinc, absorbance, IonPac CS5A, post column, semiconductor cleaning bath, t-metals, Transition Metals, Visible Absorbance, semiconductor, Nickel, TN 73892, TN73892, AN 131 |
| Matrix: | Semiconductor cleaning bath |
| Author: | Mark Laikthman and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.