Find methods for your needs
AN87: Determination of Sugar Alcohols in Confections and Fruit Juices by HPAE-PAD
Description
Sugar alcohols are used in confectionary products because they impart a sweet taste without the calories associated with sugars. Their use in foods is regulated due to their laxative and diuretic properties. This application note uses the Dionex CarboPac MA1 column and pulsed amperometric detection to determine sugar alcohols without the need for derivitization or postcolumn reagents.| Market: | Food and Beverage |
| Keywords: | candy, CarboPac MA1, Electochemical detection, Glucose, Hydrogenated glucose syrup, Chewing gum, Xylitol, Dulcitol, Myo-inositol, Laxitive, Diruetic, Glycerol, Sorbitol, apple juice, mannitol, PAD, fructose, TN 20, TN 70671, TN20, TN70671, AN 87 |
| Matrix: | Candy, Juice |
| Affiliation: | Dionex Corporation |
For Research Use Only. Not for use in diagnostic procedures.