Find methods for your needs
Detailed profiles of ethoxylated lauryl sulfates by HPLC-CAD
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of ethoxylated laurly sulfates. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.Market: | Clinical Research |
Keywords: | Acclaim Surfactant Plus, Charged aerosol detection, Ethoxylated lauryl sulfates, HPLC, POE(2), POE(30), Surfactant |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.