Find methods for your needs
Improved analysis of liquid hand soap using a Thermo Scientific Acclaim Surfactant Plus column with charged aerosol detection Be the first to rate this application
improved analysis liquid hand soap using a thermo scientific acclaim surfactant plus column with charged aerosol detection
# Compound Name Compound Class RT (min)
1 Xylene sulfonate Surfactant 3.89
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of liquid hand soap. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.
Market: Clinical Research
Keywords: Acclaim Surfactant Plus, CAD, Charged aerosol detection, Hand soap, HPLC, Surfactant, Xylene sulfonate
Matrix: Hand soap
Uploaded on 3/4/2015.
For Research Use Only. Not for use in diagnostic procedures.