Find methods for your needs
Quick separation of triclosan in toothpaste using a Thermo Scientific Acclaim RSLC C8 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of triclosan in toothpaste. The separation is performed on a Thermo Scientific Acclaim C8 RSLC column, with UV detection at 218 nm.| Market: | Clinical Research |
| Keywords: | Acclaim C8, HPLC, Saccharin, Toothpaste, Triclosan, UV detection |
| Matrix: | Toothpaste |
For Research Use Only. Not for use in diagnostic procedures.