Find methods for your needs
Improved analysis of fat-soluble vitamins and carotenoids in a vitamin tablet using a Thermo Scientific Acclaim PolarAdvantage (PA) column
Description
The Thermo Scientific HPLC system is applied for the analysis of fat-soluble vitamins and carotenoids in a vitamin tablet. The separation is performed using a Thermo Scientific Acclaim PolarAdvantage (PA) column, with UV detection at 450 and 285 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage, Carotene, Carotenoids, Fat-soluble vitamins, HPLC, Lutein, Lycopene, Retinol, Tablet, Tocopherol, UV detection, Vitamins |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.