Find methods for your needs
AN161: Determination of Metal Cyanide Complexes by Ion Chromatography with On-Line Sample Preconcentration and UV Absorbance Detection.
Description
In this application note, an improved IC method is described for the determination of metal cyanide complexes in environmental waters. The metal cyanide complexes of silver, gold, copper, nickel, iron, and cobalt are separated on an anion-exchange column and quantified by measuring their absorbance at 215 nm. Sensitivity for most of the metal cyanide complexes is improved by over two orders of magnitude, compared to a direct injection, by preconcentrating metal cyanide complexes from a large sample volume onto a trap column before separation.| Market: | Environmental |
| Keywords: | Drinking Water, IonPac AS11, IonPac ATC-3 column, Metal Cyanide, Metal Cyanide Complexes, On-Line Sample Preconcentration, Potassium dicyanoargentate, Potassium dicyanoaurate, Potassium ferrocyanide (II) trihydrate, Potassium hexacyanocobaltate (III), Potassium tetracyanonickelate (II) hydrate, Preconcentration, Reagent water, Surface water, UV Absorbance |
| Matrix: | Drinking water, Reagent water, Surface water |
| Author: | David H. Thomas and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.