Find methods for your needs
AN277: Fast and Sensitive Determination of Transition Metals in Power Industry Waters Using Ion Chromatography
Description
This study describes the separation of iron (III), copper (II), nickel (II), and zinc (II) complexes with PDCA followed by postcolumn PAR reaction and absorbance detection at 530 nm. Surrogate samples include spiked deionized (DI) water (Boiling Water Reactor (BWR) surrogate) and spiked borated water containing lithium hydroxide with or without added zinc (Pressurized Water Reactor (PWR) surrogates).| Market: | Water Analysis |
| Keywords: | Borated water, Copper, CS5A, IGSCC, Iron, MetPac, Nickel, Nuclear Power Plant, PAR, PCR, PDCA, Pressurized water reactors, Trace Cation Concentrator, VWD, Zinc |
| Matrix: | Nuclear power plant water surrogates |
| Author: | Rich Kornfeld and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.