Find methods for your needs
Improved peptide mapping of ovalbumin using a Thermo Scientific Acclaim 300 C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the peptide mapping of ovalbumin. The separation is performed on a Thermo Scientific Acclaim 300 C18 HPLC column, with UV detection at 214 nm.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, HPLC, Ovalbumin, Peptide mapping, UV detection |
| Matrix: | Ovalbumin (tryptic digest) |
For Research Use Only. Not for use in diagnostic procedures.