Find methods for your needs
AN208: Determination of Bromate in Bottled Mineral Water Using the CRD 300 Carbonate Removal Device
Description
This application note describes the use of the Thermo Scientific Dionex CRD 300 Carbonate Removal Device to remove the majority of carbonate from the eluent and allow hydroxide-like performance and detection sensitivity. This device was used with the Dionex IonPac AS23 to determine bromate in bottled mineral water samples. Detection sensitivity when using the Dionex CRD 300 Carbonate Removal Device was improved compared to chromatography without the Dionex CRD 300 device.| Market: | Water Analysis |
| Keywords: | Bottled water, Bromide, Dionex AN184, Drinking water, IonPac AS23, Ozonation, Ozone, RFIC |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.