Find methods for your needs
AN273: Higher Resolution Separation of Organic Acids and Common Inorganic Anions in Wine
Description
The method described here shows a higher resolution separation of the organic acids and inorganic acids found in wine than previously reported. This approach is applied to the separation and detection of organic acids and inorganic anions in four red and white wine samples.| Market: | Food and Beverage |
| Keywords: | Chloride, Citrate, Formate, Lactate, Malate, Nitrate, OmniPac PAX-500, Oxalate, Phosphate, Shikimate, Succinate, Sulfate, Tartrate |
| Matrix: | Wine |
| Author: | C. Chantarasukon, S. Tukkeeree, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.