Find methods for your needs
TN139: Determination of Anions in Fracking Flowback Water From the Marcellus Shale Using Automated Dilution and Ion Chromatography
Description
This Technical Note describes the analysis of fracking flowback solutions using ion chromatography following in-line determination of sample conductivity and automated dilution of high ionic strength samples.| Market: | Environmental |
| Keywords: | Acetate, Bromide, Chloride, Dionex AS-AP Autosampler, Dionex IonPac AS18, Fluoride, Formate, Hydraulic Fracturing, In-line Conductivity, RFIC, Sulfate |
| Matrix: | Wastewater |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.