Find methods for your needs
AN1113 Determination of Chloride and Sulfate using Hydroxide Eluent
Description
In this study, the determination of chloride and sulfate in soil and water samples is demonstrated using two systems: an integrated Dionex ICS-2100 system that operates as a Reagent-Free IC (RFIC) system and a Dionex ICS-5000+ HPIC system that is capable of supporting high-pressure IC and is also a RFIC system. The separation is achieved using a Dionex IonPac AS22 Analytical column or, when using high-pressure IC, a Dionex IonPac AS18-4µm Analytical column.For both methods, the separated analytes are detected using suppressed conductivity detection. An ICS-6000 can be used for this application| Market: | Environmental |
| Keywords: | Dionex IonPac AS18-4μm Column, Dionex OnGuard II P Cartridges, Humic Acid, Ion Chromatography (IC), soil, suppressed conductivity detection, ICS-6000 |
| Matrix: | Soil, Water |
| Author: | Manali Aggrawal and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.