Find methods for your needs
AN1113: Determination of Chloride and Sulfate using Carbonate Eluent Be the first to rate this application
Description
In this study, the determination of chloride and sulfate in soil and water samples is demonstrated using two systems: an integrated Dionex ICS-2100 system that operates as a Reagent-Free IC (RFIC) system and a Dionex ICS-5000+ HPIC system that is capable of supporting high-pressure IC and is also a RFIC system. The separation is achieved using a Dionex IonPac AS22 Analytical column or, when using high-pressure IC, a IonPac AS18-4µm Analytical column. For both methods, the separated analytes are detected using suppressed conductivity detection. An ICS-6000 can be used for this application.
Market: Environmental
Keywords: Dionex™ OnGuard™ II P Cartridges, Humic Acid, Ion Chromatography (IC), soil, suppressed conductivity detection, Dionex IonPac AS22 column, ICS-6000
Matrix: Soil, Water
Author: Manali Aggrawal and Jeff Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 1/8/2015.
For Research Use Only. Not for use in diagnostic procedures.