Find methods for your needs
AN157: Comparison of Suppressed to Nonsuppressed Conductivity Detection for the Determination of Common Inorganic Cations
Description
This application compares suppressed to nonsuppressed conductivity detection for the determination of inorganic cations. The Thermo Scientific Dionex IonPac CS16 was used to demonstrate the capabilities of a suppressed cation system, in terms of capacity, linearity, detection limits, and typical baseline noise using a self-regenerating suppressor. A silica-based cation-exchange column, the Thermo Scientific Dionex IonPac SCS 1, was evaluated for nonsuppressed cations and the results were compared to the suppressed system.| Market: | Water Analysis |
| Keywords: | Nonsuppressed conductivity, Suppressed conductivity, Thermo Scientific Dionex IonPac CS16 Column, Thermo Scientific Dionex IonPac SCS-1 Column |
| Matrix: | Water |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.