Find methods for your needs
AN152: Determination of Sodium at the Parts-Per-Trillion Level in the Presence of High Concentrations of Ethanolamine in Power Plant Waters
Description
This application describes an IC method for the determination of trace sodium in power plant matrices (e.g., high ethanolamine matrices). The method includes a sample preconcentration column to allow for the quantification of sodium at the ng/L levels. The analytes are separated on a high-capacity Thermo Scientific Dionex IonPac CS16 column using on-line electrically generated MSA and detected using suppressed conductivity.| Market: | Other |
| Keywords: | Inorganic Cations, Sodium, Water analysis, TN 73892, TN73892, AN 152 |
| Matrix: | Ethanolamine |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.