Find methods for your needs
A rapid separation of bases using a core enhanced technology HPLC column
Description
The Thermo Scientific HPLC system is applied for the separation of basic compounds. The separation was performed using a Thermo Scientific Accucore RP-MS column in under 3 minutes, with UV detection at 215nm.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Acenaphthene, Amitriptyline, Bases, Butyl paraben, HPLC, UV detection, Propranolol, Uracil, Napthalene |
| Matrix: | N/A |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.