Find methods for your needs
Rapid determination of Pregabalin in human plasma by SPE-LC-MS/MS using Thermo Scientific SOLA SCX mixed mode SPE cartridges and a Thermo Scientific Accucore PFP HPLC column
Description
A simple, rapid and sensitive procedure for the determination of pregabalin in human plasma by LC-MS/MS was developed and evaluated. The drug was isolated from a plasma matrix using Thermo Scientific SOLA SCX solid phase extraction material, and the components of the resultant extracts were separated on a Thermo Scientific Accucore PFP HPLC column. Detection was performed on a triple quadrupole mass spectrometer. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore PFP, Gabapentin, LC-MS/MS, Pregabalin, SOLA SCX, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.