Find methods for your needs
Determination of glimepiride in formulation using a polar endcapped UHPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the determination of glimepiride in tablet formulation. The Thermo Scientific Hypersil GOLD aQ UHPLC column separates glimepiride from the excipients present in the formulation in less than 11 minutes.| Market: | Pharma |
| Keywords: | Antidiabetic, Glimepiride, Hypersil GOLD aQ, UV detection, UHPLC |
| Matrix: | Tablet |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.